N-Boc-D-prolinal
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1-Pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, (2R)-, tert-Butyl (R)-2-Formylpyrrolidine-1-carboxylate, (R)-N-BOC-Prolinal, (R)-2-Formylpyrrolidine-1-carboxylic acid tert-Butyl ester, N-Boc-D-Prolinal, N-Boc-D-prolinal, N-(tert-Butoxycarbonyl)-D-prolinal, (R)-tert-butyl 2-formylpyrrolidine-1-carboxylate
- CAS Number:73365-02-3
- Formula:C₁₀H₁₇NO₃
- MDL Number:MFCD01321389
- Molecular Weight:199.25 g/mol
- Storage Temperature:Ambient
4 Options Available Below
Product Details & Documents
Synonyms: 1-Pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, (2R)-, tert-Butyl (R)-2-Formylpyrrolidine-1-carboxylate, (R)-N-BOC-Prolinal, (R)-2-Formylpyrrolidine-1-carboxylic acid tert-Butyl ester, N-Boc-D-Prolinal, N-Boc-D-prolinal, N-(tert-Butoxycarbonyl)-D-prolinal, (R)-tert-butyl 2-formylpyrrolidine-1-carboxylate
- CAS Number:73365-02-3
- Formula:C₁₀H₁₇NO₃
- MDL Number:MFCD01321389
- Molecular Weight:199.25 g/mol
- Storage Temperature:Ambient



