Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Dibromoadamantane

1,3-Dibromoadamantane

Supplier:   Apollo Scientific
1,3-Dibromoadamantane
Supplier:  Apollo Scientific
Certificates
1,3-Dibromoadamantane

6 Options Available Below

Product Details & Documents
  • CAS Number:876-53-9
  • Formula:C₁₀H₁₄Br₂
  • MDL Number:MFCD00077218
  • Molecular Weight:294.03 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 31,90 €
    31,90 €
    1 * 25 g
       31,90 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 76,70 €
    76,70 €
    1 * 100 g
       76,70 €

  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 356,00 €
    356,00 €
    1 * 500 g
       356,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 1 770,00 €
    1 770,00 €
    1 * 2,5 kg
       1 770,00 €

    Specifications:
    New Product
    New Product