Order Entry
Export
ContactUsLinkComponent
1,4-Bis(diphenylphosphino)butane 97%
1,4-Bis(diphenylphosphino)butane 97%
Catalog # APOSOR315705-100G
Supplier:  Apollo Scientific
CAS Number:  
1,4-Bis(diphenylphosphino)butane 97%
Catalog # APOSOR315705-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :426.48 g/mol
Melting Pt:132…133.5 °C
Storage Temperature:Ambient
CAS Number:7688-25-7
Formula:C₂₈H₂₈P₂
MDL Number:MFCD00003051