0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Tris(4-methoxyphenyl)phosphine
Tris(4-methoxyphenyl)phosphine 95%
Tris(4-methoxyphenyl)phosphine 95%
Tris(4-methoxyphenyl)phosphine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Tris(4-methoxyphenyl)phosphine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
855-38-9
:
Formula:
C₂₁H₂₁O₃P
MDL Number:
MFCD00014896
:
352.37 g/mol
Melting Pt:
131…132 °C
Storage Temperature:
Ambient