Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(R,R)-(-)-2,4-Pentanediol
(R,R)-(-)-2,4-Pentanediol 98%
(R,R)-(-)-2,4-Pentanediol 98%
APOSOR315299-250MG
:
Apollo Scientific
:
(R,R)-(-)-2,4-Pentanediol 98%
APOSOR315299-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
104.15 g/mol
:
Ambient
:
42075-32-1
:
C₅H₁₂O₂
:
MFCD00063893