Order Entry
Export
ContactUsLinkComponent
(S,S)-(+)-2,4-Pentanediol 98%
(S,S)-(+)-2,4-Pentanediol 98%
Catalog # APOSOR315298-1G
Supplier:  Apollo Scientific
CAS Number:  
(S,S)-(+)-2,4-Pentanediol 98%
Catalog # APOSOR315298-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :104.15 g/mol
Storage Temperature:Ambient
CAS Number:72345-23-4
Formula:C₅H₁₂O₂
MDL Number:MFCD00064276