Order Entry
Export
ContactUsLinkComponent
(S)-(+)-1,3-Butanediol 97%
(S)-(+)-1,3-Butanediol 97%
Catalog # APOSOR315271-5G
Supplier:  Apollo Scientific
CAS Number:  
(S)-(+)-1,3-Butanediol 97%
Catalog # APOSOR315271-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :90.12 g/mol
Storage Temperature:Ambient
CAS Number:24621-61-2
Formula:C₄H₁₀O₂
MDL Number:MFCD00064278