Order Entry
Export
ContactUsLinkComponent
2,4-Dibromo-5-nitropyridine 98%
Irritant
2,4-Dibromo-5-nitropyridine 98%
Catalog # APOSOR315206-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,4-Dibromo-5-nitropyridine 98%
Catalog # APOSOR315206-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :281.8900 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:4487-57-4
Formula:C₅H₂Br₂N₂O₂
MDL Number:MFCD09832226