
2,2'-Bipyrimidyl
Catalog # APOSOR315153-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:158.16 g/mol
Melting Pt:112…116 °C
EINECS:252-137-2
:GHS07
Storage Temperature:Ambient
CAS Number:34671-83-5
Formula:C₈H₆N₄
MDL Number:MFCD00014600