Order Entry
Export
Orders LinkContactUsLinkComponent
1,1'-Dibenzoylferrocene

1,1'-Dibenzoylferrocene

New Product
Supplier:   Apollo Scientific
1,1'-Dibenzoylferrocene
Supplier:  Apollo Scientific
New Product
Certificates
1,1'-Dibenzoylferrocene

5 Options Available Below

Product Details & Documents
Synonyms:  Di(benzoylcyclopentadienyl) Iron, Bis(benzoylcyclopentadienyl) Iron
  • CAS Number:12180-80-2
  • Formula:C₂₄H₁₀FeO₂
  • MDL Number:MFCD00045455
  • Molecular Weight:386.18 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 84,00 €
    84,00 €
    1 * 25 g
       84,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 310,00 €
    310,00 €
    1 * 100 g
       310,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 490,00 €
    1 490,00 €
    1 * 500 g
       1 490,00 €

    Specifications:
    New Product
    New Product