Order Entry
Export
Orders LinkContactUsLinkComponent
 

Pyruvic acid 98%

Corrosive
Supplier:   Apollo Scientific
Pyruvic acid 98%
Supplier:  Apollo Scientific
Certificates
Pyruvic acid 98%

4 Options Available Below

Product Details & Documents
Synonyms:  α-Ketopropionic acid, 2-Oxopropionic acid, 2-Ketopropionic acid, 2-Oxopropanoic acid
  • CAS Number:127-17-3
  • Formula:C₃H₄O₃
  • MDL Number:MFCD00002585
  • Molecular Weight:88.06 g/mol
  • Boiling Pt:164…166 °C
  • Melting Pt:44541 °C
  • Density:1.267 g/cm³ (25 °C)
  • Flash Pt:83 °C
  • Storage Temperature:Refrigerator
  • EINECS:204-824-3
  • UN:3265
  • ADR:8,II

 

Product Family Options

Product Information