
(±)-trans-1,2-Diaminocyclohexane 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1121-22-8
:
Formula:C₆H₁₄N₂
MDL Number:MFCD00063747
:114.19 g/mol
Melting Pt:14…15 °C
Density:0.951 g/cm³ (25 °C)
Flash Pt:68 °C
Storage Temperature:Ambient
UN:2735
ADR:8,II
:GHS05