Order Entry
Export
ContactUsLinkComponent
5-Bromo-2,3-diaminopyridine
Irritant
5-Bromo-2,3-diaminopyridine
Catalog # APOSOR3145-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Bromo-2,3-diaminopyridine
Catalog # APOSOR3145-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :188.03 g/mol
Melting Pt:155…161 °C (dec.)
EINECS:254-172-9
 :GHS07
Storage Temperature:Ambient
CAS Number:38875-53-5
Formula:C₅H₆BrN₃
MDL Number:MFCD00460094