Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,3-Dimethylindoline

Irritant
Supplier:   Apollo Scientific
3,3-Dimethylindoline
Supplier:  Apollo Scientific
Certificates
3,3-Dimethylindoline

5 Options Available Below

Product Details & Documents
  • CAS Number:1914-02-9
  • Formula:C₁₀H₁₃N
  • MDL Number:MFCD11847210
  • Molecular Weight:147.22 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 21,80 €
    21,80 €
    1 * 100 mg
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 30,40 €
    30,40 €
    1 * 250 mg
       30,40 €

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 99,90 €
    99,90 €
    1 * 1 g
       99,90 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 491,00 €
    491,00 €
    1 * 5 g
       491,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 2 440,00 €
    2 440,00 €
    1 * 25 g
       2 440,00 €

    Specifications:
    New Product
    New Product