Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dimethyl cubane-1,4-dicarboxylate
Dimethyl Cubane-1,4-dicarboxylate 95%
Dimethyl Cubane-1,4-dicarboxylate 95%
Irritant
Dimethyl Cubane-1,4-dicarboxylate 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Dimethyl Cubane-1,4-dicarboxylate 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
29412-62-2
:
MFCD00192075
Formula:
C₁₂H₁₂O₄
MDL Number:
MFCD29060101
:
220.22 g/mol
Melting Pt:
163 °C
:
GHS07
Storage Temperature:
Refrigerator