
2-Oxa-spiro[3.3]heptan-6-one
Catalog # APOSOR312273-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:112.13 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1339892-66-8
Formula:C₆H₈O₂
MDL Number:MFCD22415239