0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2-Oxaspiro[3.5]nonan-7-one
2-Oxaspiro[3.5]nonan-7-one
2-Oxaspiro[3.5]nonan-7-one
Irritant
2-Oxaspiro[3.5]nonan-7-one
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Oxaspiro[3.5]nonan-7-one
Supplier:
Apollo Scientific
About this item
CAS Number:
1256546-74-3
:
Formula:
C₈H₁₂O₂
MDL Number:
MFCD22628754
:
140.18 g/mol
:
GHS07
Storage Temperature:
Refrigerator