
7-Azaspiro[3.5]nonane
Catalog # APOSOR312259-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:125.21 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:766-34-7
Formula:C₈H₁₅N
MDL Number:MFCD14581275