4-Nitro-1,3-dihydro-2H-benzimidazol-2-one
Catalog # APOSOR311139-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:179.14 g/mol
Storage Temperature:Ambient
CAS Number:85330-50-3
Formula:C₇H₅N₃O₃
MDL Number:MFCD09835484