Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,4-Dichlorobenzhydrazide

Irritant
New Product
Supplier:   Apollo Scientific
2,4-Dichlorobenzhydrazide
Supplier:  Apollo Scientific
New Product
Certificates
2,4-Dichlorobenzhydrazide

2 Options Available Below

Product Details & Documents
Synonyms:  2,4-Dichlorobenzoic hydrazide
  • CAS Number:5814-06-2
  • Formula:C₇H₆Cl₂N₂O
  • MDL Number:MFCD00025113
  • Molecular Weight:205.04 g/mol
  • Melting Pt:168…170 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 72,40 €
    72,40 €
    1 * 1 g
       72,40 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 808,00 €
    808,00 €
    1 * 25 g
       808,00 €

    Specifications:
    New Product
    New Product