Order Entry
Export
ContactUsLinkComponent
2-Methyl-11-oxo-11H-pyrido[2,1-b]quinazoline-6-carboxylic acid
2-Methyl-11-oxo-11H-pyrido[2,1-b]quinazoline-6-carboxylic acid
Catalog # APOSOR310763-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Methyl-11-oxo-11H-pyrido[2,1-b]quinazoline-6-carboxylic acid
Catalog # APOSOR310763-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :254.25 g/mol
Storage Temperature:Ambient
CAS Number:1111000-64-6
Formula:C₁₄H₁₀N₂O₃
MDL Number:MFCD20754075