Order Entry
Export
ContactUsLinkComponent
[7-(3-Methyl-1,2,4-oxadiazol-5-yl)[1,2,4]triazolo[4,3-a]pyridin-3-yl]acetic acid
[7-(3-Methyl-1,2,4-oxadiazol-5-yl)[1,2,4]triazolo[4,3-a]pyridin-3-yl]acetic acid
Catalog # APOSOR310678-1G
Supplier:  Apollo Scientific
CAS Number:  
[7-(3-Methyl-1,2,4-oxadiazol-5-yl)[1,2,4]triazolo[4,3-a]pyridin-3-yl]acetic acid
Catalog # APOSOR310678-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :259.23 g/mol
Storage Temperature:Ambient
CAS Number:1338661-82-7
Formula:C₁₁H₉N₅O₃
MDL Number:MFCD20528959