1,3-Diethynylbenzene 97%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:1785-61-1
- Formula:C₁₀H₆
- MDL Number:MFCD00041686
- Molecular Weight:126.16 g/mol
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
- CAS Number:1785-61-1
- Formula:C₁₀H₆
- MDL Number:MFCD00041686
- Molecular Weight:126.16 g/mol
- Storage Temperature:Ambient



