
2,5-Dichloro-2,5-dimethylhexane 98%
Catalog # APOSOR310336-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:183.12 g/mol
Boiling Pt:180 °C
Melting Pt:68 °C
:GHS07
Storage Temperature:Ambient
CAS Number:6223-78-5
Formula:C₈H₁₆Cl₂
MDL Number:MFCD00126854