Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,7-Dibromo-9,9-dipropylfluorene
2,7-Dibromo-9,9-dipropylfluorene
2,7-Dibromo-9,9-dipropylfluorene
2,7-Dibromo-9,9-dipropylfluorene
Supplier:
Apollo Scientific
Certificates
Expand image
2,7-Dibromo-9,9-dipropylfluorene
Supplier:
Apollo Scientific
About this item
CAS Number:
157771-56-7
:
Formula:
C₁₉H₂₀Br₂
MDL Number:
MFCD12024280
:
408.18 g/mol
Storage Temperature:
Ambient