
Methyl-5-aminosalicylate
Catalog # APOSOR31020-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:167.1620 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:42753-75-3
Formula:C₈H₉NO₃
MDL Number:MFCD01317557