Order Entry
Export
ContactUsLinkComponent
2-Amino-5-iodo-4,6-dimethylpyridine 95%
Irritant
2-Amino-5-iodo-4,6-dimethylpyridine 95%
Catalog # APOSOR310150-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Amino-5-iodo-4,6-dimethylpyridine 95%
Catalog # APOSOR310150-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :248.07 g/mol
Melting Pt:164…166 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:885952-12-5
Formula:C₇H₉IN₂
MDL Number:MFCD06798226