N-(5-Iodo-3-methyl-pyridin-2-yl)-2,2-dimethylpropionamide
Catalog # APOSOR310100-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:318.16 g/mol
Storage Temperature:Ambient
CAS Number:677327-29-6
Formula:C₁₁H₁₅IN₂O
MDL Number:MFCD04218302