Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromoimidazo[1,2-b]pyridazine
3-Bromoimidazo[1,2-b]pyridazine 98%
3-Bromoimidazo[1,2-b]pyridazine 98%
:
Apollo Scientific
3-Bromoimidazo[1,2-b]pyridazine 98%
:
Apollo Scientific
:
18087-73-5
:
:
C6H4BrN3
:
MFCD09757672
:
198.0201 g/mol
:
146…149 °C
:
GHS07
:
Ambient