
2-Ethyl-1,3-benzoxazol-5-amine
Catalog # APOSOR30966-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:GHS07
Storage Temperature:Ambient
CAS Number:204771-75-5
Formula:C₉H₁₀N₂O