Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-4,6-dimethylpyridine
2-Chloro-4,6-dimethylpyridine 98%
2-Chloro-4,6-dimethylpyridine 98%
Irritant
2-Chloro-4,6-dimethylpyridine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Chloro-4,6-dimethylpyridine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
30838-93-8
:
Formula:
C₇H₈ClN
MDL Number:
MFCD08277279
:
141.60 g/mol
:
GHS07
Storage Temperature:
Ambient