tert-Butyl-3-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate
Catalog # APOSOR308119-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:254.33 g/mol
Storage Temperature:Ambient
CAS Number:169206-67-1
Formula:C₁₃H₂₂N₂O₃
MDL Number:MFCD11111226