0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
5-Chloro-1H-pyrrolo[2,3-b]pyridine
5-Chloro-1H-pyrrolo[2,3-b]pyridine
5-Chloro-1H-pyrrolo[2,3-b]pyridine
Corrosive
Irritant
5-Chloro-1H-pyrrolo[2,3-b]pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Irritant
5-Chloro-1H-pyrrolo[2,3-b]pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
866546-07-8
:
Formula:
C₇H₅ClN₂
MDL Number:
MFCD06659676
:
152.58 g/mol
Melting Pt:
161…165 °C
:
GHS05
:
GHS07
Storage Temperature:
Ambient