0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
N-Boc-N-methyl-1,3-diaminopropane
N-Boc-N-methyl-1,3-diaminopropane
N-Boc-N-methyl-1,3-diaminopropane
Irritant
N-Boc-N-methyl-1,3-diaminopropane
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
N-Boc-N-methyl-1,3-diaminopropane
Supplier:
Apollo Scientific
About this item
CAS Number:
150349-36-3
:
Formula:
C₉H₂₀N₂O₂
MDL Number:
MFCD01631189
:
188.271 g/mol
:
GHS07
Storage Temperature:
Ambient