5,7-Dichloro-2-methylquinoline
Catalog # APOSOR307984-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:26933-09-5
Formula:C₁₀H₇Cl₂N