Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,8-Dichloro-2-methylquinoline
4,8-Dichloro-2-methylquinoline 97%
4,8-Dichloro-2-methylquinoline 97%
Corrosive
Toxic
4,8-Dichloro-2-methylquinoline 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Toxic
4,8-Dichloro-2-methylquinoline 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
75896-69-4
:
Formula:
C₁₀H₇Cl₂N
Melting Pt:
86…87 °C
Storage Temperature:
Ambient
UN:
2811
ADR:
6.1,III
:
GHS05
:
GHS06