Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-5,8-dimethylquinoline-3-carbonitrile
2-Chloro-5,8-dimethylquinoline-3-carbonitrile
2-Chloro-5,8-dimethylquinoline-3-carbonitrile
2-Chloro-5,8-dimethylquinoline-3-carbonitrile
Supplier:
Apollo Scientific
Certificates
Expand image
2-Chloro-5,8-dimethylquinoline-3-carbonitrile
Supplier:
Apollo Scientific
About this item
CAS Number:
351363-12-7
:
Formula:
C₁₂H₉ClN₂
MDL Number:
MFCD02741714
:
216.67 g/mol
Storage Temperature:
Ambient