3-(Chloromethyl)-2,7-dichloroquinoline
Catalog # APOSOR307908-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:137776-95-5
Formula:C₁₀H₆Cl₃N