
2,6-Dichloro-3-methylquinoline
Catalog # APOSOR307884-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:GHS07
Storage Temperature:Ambient
CAS Number:132118-32-2
Formula:C₁₀H₇Cl₂N