6-Bromo-4-chloro-2-phenylquinoline
Catalog # APOSOR307875-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:860195-69-3
Formula:C₁₅H₉BrClN
MDL Number:MFCD02684146