
4,6-Dichloropicolinic acid
Catalog # APOSOR30786-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:192.0000 g/mol
Melting Pt:111…114 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:88912-25-8
Formula:C₆H₃Cl₂NO₂
MDL Number:MFCD09910267