Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dichloro-2-methylquinoline
4,6-Dichloro-2-methylquinoline
4,6-Dichloro-2-methylquinoline
4,6-Dichloro-2-methylquinoline
Supplier:
Apollo Scientific
Certificates
Expand image
4,6-Dichloro-2-methylquinoline
Supplier:
Apollo Scientific
About this item
CAS Number:
53342-53-3
:
Formula:
C₁₀H₇Cl₂N
MDL Number:
MFCD00272372
Storage Temperature:
Ambient