6-Oxo-1,6-dihydropyridazine-4-carboxylic acid 95+%
Catalog # APOSOR30774-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:140.0969 g/mol
Storage Temperature:Ambient
CAS Number:867130-58-3
Formula:C₅H₄N₂O₃
MDL Number:MFCD08437486