Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dichloroquinoline
2,6-Dichloroquinoline
2,6-Dichloroquinoline
:
Apollo Scientific
2,6-Dichloroquinoline
:
Apollo Scientific
:
1810-72-6
:
:
C₉H₅Cl₂N
:
MFCD03427194
:
198.0500 g/mol
:
161…164 °C
:
GHS07
:
Ambient