Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(R)-(-)-1,2-Propanediol
(R)-(-)-1,2-Propanediol 98%
(R)-(-)-1,2-Propanediol 98%
(R)-(-)-1,2-Propanediol 98%
Supplier:
Apollo Scientific
Certificates
Expand image
(R)-(-)-1,2-Propanediol 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
4254-14-2
:
Formula:
C₃H₈O₂
MDL Number:
MFCD00066248
:
76.1 g/mol
Storage Temperature:
Ambient