Order Entry
Export
ContactUsLinkComponent
4,6-Dichloro-2-(methylthio)-5-nitropyrimidine 98%
4,6-Dichloro-2-(methylthio)-5-nitropyrimidine 98%
Catalog # APOSOR307014-1G
Supplier:  Apollo Scientific
CAS Number:  
4,6-Dichloro-2-(methylthio)-5-nitropyrimidine 98%
Catalog # APOSOR307014-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :240.0672 g/mol
Storage Temperature:Ambient
CAS Number:1979-96-0
Formula:C₅H₃Cl₂N₃O₂S
MDL Number:MFCD02323208