0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1H-Pyrazolo[3,4-b]pyridine
1H-Pyrazolo[3,4-b]pyridine 97%
1H-Pyrazolo[3,4-b]pyridine 97%
Irritant
1H-Pyrazolo[3,4-b]pyridine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1H-Pyrazolo[3,4-b]pyridine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
271-73-8
:
Formula:
C₆H₅N₃
MDL Number:
MFCD05663981
:
119.12 g/mol
Melting Pt:
93…95 °C
:
GHS07
Storage Temperature:
Ambient