
1H-Pyrazolo[3,4-c]pyridine 95+%
Catalog # APOSOR30697-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:119.1240 g/mol
Melting Pt:100…108 °C
:GHS07
Storage Temperature:Ambient
CAS Number:271-47-6
Formula:C₆H₅N₃
MDL Number:MFCD00661308