Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-3-chloroimidazo[1,2-a]pyridine
2-Bromo-3-chloroimidazo[1,2-a]pyridine
2-Bromo-3-chloroimidazo[1,2-a]pyridine
2-Bromo-3-chloroimidazo[1,2-a]pyridine
Supplier:
Apollo Scientific
Certificates
Expand image
2-Bromo-3-chloroimidazo[1,2-a]pyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
1159511-19-9
:
Formula:
C₇H₄BrClN₂
MDL Number:
MFCD11869753
:
231.4800 g/mol
Storage Temperature:
Ambient