Order Entry
Export
Orders LinkContactUsLinkComponent
2,6-Dimethylhydroquinone

2,6-Dimethylhydroquinone

Toxic
Health
Supplier:   Apollo Scientific
2,6-Dimethylhydroquinone
Supplier:  Apollo Scientific
Certificates
2,6-Dimethylhydroquinone

5 Options Available Below

Product Details & Documents
Synonyms:  2,5-Dihydroxy-m-xylene
  • CAS Number:654-42-2
  • Formula:C₈H₁₀O₂
  • MDL Number:MFCD00016466
  • Molecular Weight:138.17 g/mol
  • Melting Pt:154 °C
  • Storage Temperature:Ambient
  • EINECS:211-505-2
  • UN:2811
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 21,80 €
    21,80 €
    1 * 250 mg
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 24,60 €
    24,60 €
    1 * 1 g
       24,60 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 46,40 €
    46,40 €
    1 * 5 g
       46,40 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 222,00 €
    222,00 €
    1 * 25 g
       222,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 883,00 €
    883,00 €
    1 * 100 g
       883,00 €

    Specifications:
    New Product
    New Product